Y0001232
Fosinopril impurity I
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
[(RS)-[(1SR)-2-Methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphoryl]acetic acid
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
fosinopril
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
[P](=O)(OC(OC(=O)CC)C(C)C)(CCCCc1ccccc1)CC(=O)O
InChI
1S/C19H29O6P/c1-4-18(22)24-19(15(2)3)25-26(23,14-17(20)21)13-9-8-12-16-10-6-5-7-11-16/h5-7,10-11,15,19H,4,8-9,12-14H2,1-3H3,(H,20,21)
InChI key
BGHVPSAAFKIBID-UHFFFAOYSA-N
Related Categories
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
Other Notes
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service