Skip to Content
Merck
All Photos(1)

Key Documents

Y0001101

Interferon β-1a

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

interferon

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

dry ice

storage temp.

−70°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Interferon β-1a EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Huang et al.
Journal of neuroimmunology, 258(1-2), 27-31 (2013-03-16)
The effect of pharmacologically relevant doses of interferon (IFN) β-1a on B-cell expression of B7.1 and B7.2 was investigated. Culture of peripheral blood mononuclear cells with IFN β-1a 100 IU/mL decreased B-cell expression of B7.1 and increased B7.2 expression. Interleukin-10
N N Spirin et al.
Zhurnal nevrologii i psikhiatrii imeni S.S. Korsakova, 112(8), 27-33 (2012-10-26)
Authors have followed up 230 patients with multiple sclerosis treated with disease modifying drugs (DMD) using the data of the Multiple sclerosis register of the Yaroslavl oblast during 2009-2011. Original drugs and their generics registered in Russia are used. Patients
Gábor Széplaki et al.
Ideggyogyaszati szemle, 65(11-12), 369-376 (2013-01-08)
Fingolimod is a sphingosine-1 phosphate receptor modulator, which is effective in the treatment of severe relapsing-remitting form of multiple sclerosis. Once daily oral use of fingolimod decreased the annualized relapse rate, inflammatory brain lesion activity and the rate of brain
Alemtuzumab for multiple sclerosis: who and when to treat?
Till Sprenger et al.
Lancet (London, England), 380(9856), 1795-1797 (2012-11-07)
Richard A C Hughes et al.
The Cochrane database of systematic reviews, 2(2), CD008630-CD008630 (2013-03-02)
Plasma exchange and intravenous immunoglobulin, but not corticosteroids, are beneficial in Guillain-Barré syndrome (GBS). The efficacy of other pharmacological agents is unknown. This review was first published in 2011 and this update in 2013. To review systematically the evidence from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service