Skip to Content
Merck
All Photos(1)

Key Documents

Y0000733

Dacarbazine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-(3,3-Dimethyl-1-triazenyl)imidazole-4-carboxamide, DTIC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10N6O
CAS Number:
Molecular Weight:
182.18
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dacarbazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN(C)\N=N\c1[nH]cnc1C(N)=O

InChI

1S/C6H10N6O/c1-12(2)11-10-6-4(5(7)13)8-3-9-6/h3H,1-2H3,(H2,7,13)(H,8,9)/b11-10+

InChI key

FDKXTQMXEQVLRF-ZHACJKMWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dacarbazine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annette E Hay et al.
Hematology/oncology clinics of North America, 28(1), 49-63 (2013-11-30)
Because long-term survival of patients with nonbulky stage IA to IIA Hodgkin lymphoma is dependent on disease control and avoidance of late toxic effects associated with the treatment received, the initial choice of treatment can be associated with trade-offs that
O Mizuno
Nihon Naibunpi Gakkai zasshi, 70(9), 1039-1046 (1994-11-20)
A 49-year-old woman had a right adrenalectomy for pheochromocytoma in April 1989. In May 1990 she underwent an operation to remove paraaortic lymph nodes, and the lymph nodes showed pheochromocytoma. Twenty-two months after the first operation, metastases to the left
[Dacarbazine].
M D'incan et al.
Annales de dermatologie et de venereologie, 128(4), 517-525 (2001-06-08)
Kamila Koprowska et al.
Postepy higieny i medycyny doswiadczalnej (Online), 65, 734-751 (2011-12-17)
Melanoma is a tumour derived from melanocytes, cells of neuroectodermal origin. Melanoma treatment represents a challenge to oncologists due to its aggressive course and early and multiple metastases. Surgical excision of lesions is a highly effective intervention, but only in
Alexander M M Eggermont et al.
European journal of cancer (Oxford, England : 1990), 40(12), 1825-1836 (2004-08-04)
Since dacarbazine was approved for treating metastatic melanoma in the 1970s, numerous studies have evaluated whether different schedules and dacarbazine-based combinations improve clinical outcomes. This evidence-based review shows that combining dacarbazine with other drugs having single-agent activity and/or hormonal or

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service