Skip to Content
Merck
All Photos(1)

Key Documents

Y0000722

Codeine

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

codeine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2C[C@@H]3C4C=C[C@H](O)[C@@H]5Oc1c2[C@]45CCN3C

InChI

1S/C18H21NO3/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-/m0/s1

InChI key

OROGSEYTTFOCAN-DNJOTXNNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Codeine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheena Derry et al.
The Cochrane database of systematic reviews, (4)(4), CD008099-CD008099 (2010-04-16)
Codeine is an opioid metabolised to active analgesic compounds, including morphine. It is widely available by prescription, and combination drugs including low doses of codeine are commonly available without prescription. To assess the efficacy, the time to onset of analgesia
Marieke Nauta et al.
American journal of surgery, 198(2), 256-261 (2009-07-25)
Cesarean section, episiotomy, and third and perineal tears are associated with significant tissue damage, causing pain in the immediate postpartum period. The current standard in North America is to prescribe oral acetaminophen/codeine (A + C) for postpartum pain. Codeine has
Lauren E Kelly et al.
Therapeutic drug monitoring, 34(3), 249-256 (2012-05-10)
Codeine is an old and commonly used analgesic agent for mild to moderate pain. It is the prototypical "prodrug" in that its analgesic effect is almost wholly dependent on its biotransformation to morphine, a process that is mediated by the
Parvaz Madadi et al.
Pharmacogenomics, 9(9), 1267-1284 (2008-09-11)
Codeine has been used medicinally since the 1800s as an analgesic and antitussive agent. Although very few studies have methodically examined the safety of codeine use in the pediatric age group, it is nonetheless commonly prescribed to children and breastfeeding
Hirokazu Toubou et al.
Legal medicine (Tokyo, Japan), 16(5), 308-313 (2014-06-01)
A simple and precise drug screening method was developed for the detection of abused drugs in whole blood by using the Oratect® III device that is usually employed for the detection of drugs in saliva. Whole blood was acidified with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service