Skip to Content
Merck
All Photos(1)

Key Documents

Y0000695

Naloxone for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Naloxone hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21NO4 · HCl · 2H2O
CAS Number:
Molecular Weight:
399.87
Beilstein:
6260807
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

naloxone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.Cl.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)CC=C

InChI

1S/C19H21NO4.ClH.2H2O/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11;;;/h2-4,14,17,21,23H,1,5-10H2;1H;2*1H2/t14-,17+,18+,19-;;;/m1.../s1

InChI key

TXMZWEASFRBVKY-IOQDSZRYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naloxone for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Easy-to-use overdose antidote earns fast-track approval.
Bridget M Kuehn
JAMA, 311(16), 1600-1600 (2014-04-24)
Heroin and naloxone.
Annie Lagoy
The American journal of nursing, 114(10), 12-12 (2014-09-25)
Time for universal provision of take-home naloxone.
Tim Crocker-Buque et al.
Lancet (London, England), 384(9940), 308-308 (2014-07-30)
Heroin: life, death, and politics.
Joy Jacobson
The American journal of nursing, 114(5), 22-23 (2014-04-25)
Edmund Allen Liles et al.
Hospital practice (1995), 42(2), 70-76 (2014-04-29)
Patients with sickle cell disease (SCD) suffer from intermittent vaso-occlusive pain crises (VOCs). These crises lead to frequent hospitalizations, significant morbidity, and increased mortality risk. Care pathways can enhance efficiency and quality of care. Our study sought to evaluate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service