Skip to Content
Merck
All Photos(1)

Key Documents

Y0000515

Glimepiride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Glimepiride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34N4O5S
CAS Number:
Molecular Weight:
490.62
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glimepiride

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC1=C(C)CN(C(=O)NCCc2ccc(cc2)S(=O)(=O)NC(=O)N[C@H]3CC[C@H](C)CC3)C1=O

InChI

1S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/t16-,19-

InChI key

WIGIZIANZCJQQY-RUCARUNLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Glimepiride is currently used to treat type 2 diabetes.

Biochem/physiol Actions

Glimepiride is a potent blocker of cardiac KATP channels activated by pinacidil with an IC50 of 6.8 nM.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary T Korytkowski
Pharmacotherapy, 24(5), 606-620 (2004-05-28)
Sulfonylureas, which have evolved through two generations since their introduction nearly 50 years ago, remain the most frequently prescribed oral agents for treatment of patients with type 2 diabetes mellitus. Glyburide, glipizide, and glimepiride, the newest sulfonylureas, are as effective
T F Veneman et al.
The Netherlands journal of medicine, 52(5), 179-186 (1998-07-04)
Disturbances in insulin secretion and insulin action are both involved in the pathophysiology of type 2 (or non-insulin-dependent) diabetes mellitus. The newly developed sulfonylurea (SU) derivative glimepiride has a marked insulin secretory effect both in vitro and in vivo, and
M Nauck et al.
Diabetes, obesity & metabolism, 15(3), 204-212 (2012-09-19)
To investigate efficacy and safety of dual therapy with liraglutide and metformin in comparison to glimepiride and metformin, and metformin monotherapy over 2 years in patients with type 2 diabetes. In the 26-week the Liraglutide Effect and Action in Diabetes
H D Langtry et al.
Drugs, 55(4), 563-584 (1998-04-30)
Glimepiride is a sulphonylurea agent that stimulates insulin release from pancreatic beta-cells and may act via extrapancreatic mechanisms. It is administered once daily to patients with type 2 (non-insulin-dependent) diabetes mellitus in whom glycaemia is not controlled by diet and
Massimo Massi-Benedetti
Clinical therapeutics, 25(3), 799-816 (2003-07-11)
Sulfonylureas (SUs) have been used for many years as first-line therapy for patients with type 2 diabetes mellitus whose blood glucose levels have not been effectively controlled by diet and exercise alone. Glimepiride is a once-daily SU that was introduced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service