Skip to Content
Merck
All Photos(1)

Key Documents

Y0000467

Thiamine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Aneurine hydrochloride, Vitamin B1 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17ClN4OS · HCl
CAS Number:
Molecular Weight:
337.27
Beilstein:
3851771
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

thiamine

manufacturer/tradename

EDQM

mp

250 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1=NC(N)=C(C[N+]2=CSC(CCO)=C2C)C=N1.Cl.[Cl-]

InChI

1S/C12H17N4OS.2ClH/c1-8-11(3-4-17)18-7-16(8)6-10-5-14-9(2)15-12(10)13;;/h5,7,17H,3-4,6H2,1-2H3,(H2,13,14,15);2*1H/q+1;;/p-1

InChI key

DPJRMOMPQZCRJU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing pharmacopoeia.

Thiamine hydrochloride is a water soluble vitamin, which belongs to the B-complex group of vitamins. It can exist both as a hemihydrate and as a nonstoichiometric hydrate. It can generally act like a co-enzyme for biochemical reactions involved in carbohydrate metabolism.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ashish K Singh et al.
Alcoholism, clinical and experimental research, 38(7), 2127-2137 (2014-06-17)
Chronic ethanol (EtOH) consumption is associated with oxidative tissue damage, decrease in antioxidant enzyme activities, and increase in hepatic and plasma lipids. This study investigates the effect of modified probiotic Escherichia coli Nissle 1917 (EcN) secreting pyrroloquinoline quinone (PQQ) against
High-performance liquid chromatography method for the simultaneous determination of thiamine hydrochloride, pyridoxine hydrochloride and cyanocobalamin in pharmaceutical formulations using coulometric electrochemical and ultraviolet detection
Marszall LM, et al.
Journal of Chromatography A, 1094(1-2), 91-98 (2005)
Katherine R Heal et al.
Rapid communications in mass spectrometry : RCM, 28(22), 2398-2404 (2014-10-11)
Vitamin B(12) is an essential nutrient for more than half of surveyed marine algae species, but methods for directly measuring this important cofactor in seawater are limited. Current mass spectrometry methods do not quantify all forms of B(12), potentially missing
Qiang Zhang et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(10), 1656-1662 (2014-07-27)
The clinical development of fedratinib, a Janus kinase (JAK2) inhibitor, was terminated after reports of Wernicke's encephalopathy in myelofibrosis patients. Since Wernicke's encephalopathy is induced by thiamine deficiency, investigations were conducted to probe possible mechanisms through which fedratinib may lead
Irene Delgado-Blanca et al.
Analytical and bioanalytical chemistry, 407(2), 521-528 (2014-11-06)
A new approach for the implementation of liquid-liquid extraction in a sequential injection manifold is presented. The manifold consists of two syringe pumps and one multi-position valve. The use of a 30 % MeOH/H2O (v/v) solution as the carrier, with isobutanol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service