Skip to Content
Merck
All Photos(1)

Key Documents

Y0000358

Clofazimine for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Clofazimine, N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H22Cl2N4
CAS Number:
Molecular Weight:
473.40
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clofazimine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)\N=C1/C=C2N(c3ccc(Cl)cc3)c4ccccc4N=C2C=C1Nc5ccc(Cl)cc5

InChI

1S/C27H22Cl2N4/c1-17(2)30-24-16-27-25(15-23(24)31-20-11-7-18(28)8-12-20)32-22-5-3-4-6-26(22)33(27)21-13-9-19(29)10-14-21/h3-17,31H,1-2H3/b30-24+

InChI key

WDQPAMHFFCXSNU-BGABXYSRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clofazimine for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clofazimine.
Tuberculosis (Edinburgh, Scotland), 88(2), 96-99 (2008-05-20)
Moloko C Cholo et al.
The Journal of antimicrobial chemotherapy, 67(2), 290-298 (2011-10-25)
Clofazimine, a lipophilic riminophenazine antibiotic, possesses both antimycobacterial and anti-inflammatory activities. However, its efficacy has been demonstrated only in the treatment of leprosy, not in human tuberculosis, despite the fact that this agent is impressively active in vitro against multidrug-resistant
J C Garrelts
DICP : the annals of pharmacotherapy, 25(5), 525-531 (1991-05-01)
This article reviews the chemistry, pharmacology, spectrum of activity, pharmacokinetics, clinical efficacy in leprosy and Mycobacterium avium complex (MAC) infection, adverse effects, drug interactions, and special considerations of clofazimine. The drug is active in vivo against M. leprae and in
Clofazimine--a review.
B Kumar
Indian journal of leprosy, 63(1), 78-92 (1991-01-01)
The pharmacology, metabolism, and chemistry of clofazimine.
R O'Connor et al.
Drug metabolism reviews, 27(4), 591-614 (1995-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service