Skip to Content
Merck
All Photos(1)

Key Documents

Y0000299

Fluphenazine sulfoxide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Fluphenazine sulphoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H26F3N3O2S
CAS Number:
Molecular Weight:
453.52
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

fluphenazine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc2c(cc1)[S](=O)c3c(cccc3)N2CCCN4CCN(CC4)CCO

InChI

1S/C22H26F3N3O2S/c23-22(24,25)17-6-7-21-19(16-17)28(18-4-1-2-5-20(18)31(21)30)9-3-8-26-10-12-27(13-11-26)14-15-29/h1-2,4-7,16,29H,3,8-15H2

InChI key

UFPPOFUTIWYLNR-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluphenazine sulfoxide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K K Midha et al.
Journal of pharmacological methods, 19(1), 63-74 (1988-03-01)
Antisera to fluphenazine sulfoxide were raised in New Zealand white rabbits to an immunogen synthesized by covalent linkage of bovine serum albumin to 10-[[3-[4-(4-carboxybutyl)-1-piperazinyl] propyl]]-2-trifluoromethyl-10H-phenothiazine 5-sulfoxide. With use of an antiserum, a radioimmunoassay for fluphenazine sulfoxide was developed that is
K K Midha et al.
Psychopharmacology, 93(3), 369-373 (1987-01-01)
Highly sensitive radioimmunoassays were applied to study the sulfoxidation of fluphenazine in 30 schizophrenic patients maintained on either 5 mg or 25 mg fluphenazine decanoate by intramuscular injection every 14 days over a period of 6 months. The presence of
S R Marder et al.
The British journal of psychiatry : the journal of mental science, 158, 658-665 (1991-05-01)
The levels of fluphenazine and fluphenazine sulphoxide in schizophrenic patients who were randomly assigned to receive either 5 mg or 25 mg of fluphenazine decanoate every two weeks were monitored. Patients treated with 25 mg of fluphenazine decanoate required three
K K Midha et al.
Journal of psychiatry & neuroscience : JPN, 19(4), 254-264 (1994-07-01)
This review covers some recent work on: 1. The effects of route of administration on the pharmacokinetics of fluphenazine and some of its metabolites; 2. The clinical pharmacokinetics of fluphenazine in acute patients medicated with oral fluphenazine; 3. The clinical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service