Skip to Content
Merck
All Photos(1)

Key Documents

Y0000119

Hydroxycarbamide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hydroxyurea, Hydroxycarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CONHOH
CAS Number:
Molecular Weight:
76.05
Beilstein:
1741548
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hydroxycarbamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)NO

InChI

1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)

InChI key

VSNHCAURESNICA-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydroxycarbamide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Anti-neoplastic. Inactivates ribonucleoside reductase by forming a free radical nitroxide that binds a tyrosyl free radical in the active site of the enzyme. This blocks the synthesis of deoxynucleotides, which inhibits DNA synthesis and induces synchronization or cell death in S-phase.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pilvi Riihilä et al.
The Journal of investigative dermatology, 135(2), 579-588 (2014-09-04)
The incidence of cutaneous squamous cell carcinoma (cSCC) is rising worldwide. We have examined the role of complement components in the progression of cSCC. Analysis of cSCC cell lines (n=8) and normal human epidermal keratinocytes (n=11) with whole transcriptome profiling
John J Strouse et al.
Pediatrics, 122(6), 1332-1342 (2008-12-03)
Hydroxyurea is the only approved medication for the treatment of sickle cell disease in adults; there are no approved drugs for children. Our goal was to synthesize the published literature on the efficacy, effectiveness, and toxicity of hydroxyurea in children
A Tefferi et al.
Leukemia, 28(12), 2300-2303 (2014-05-06)
The impact of calreticulin (CALR) mutations on long-term survival in essential thrombocythemia (ET) was examined in 299 patients whose diagnosis predated 2006. Mutational frequencies were 53% for Janus kinase 2 (JAK2), 32% for CALR and 3% for MPL; the remaining
Nancy S Green et al.
Pediatric blood & cancer, 56(2), 177-181 (2010-09-11)
Fetal hemoglobin (HbF) levels are generally inversely proportional to severity of sickle cell disease (SCD) for given sickle phenotypes. Molecular regulation of HbF occurs through complex interactions cis and trans to the beta globin gene locus. Novel insights made through
Russell E Ware
Blood, 115(26), 5300-5311 (2010-03-13)
Hydroxyurea has many characteristics of an ideal drug for sickle cell anemia (SCA) and provides therapeutic benefit through multiple mechanisms of action. Over the past 25 years, substantial experience has accumulated regarding its safety and efficacy for patients with SCA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service