Skip to Content
Merck
All Photos(1)

Key Documents

Y0000113

Carbachol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Carbamoylcholine chloride, (2-Hydroxyethyl)trimethylammonium chloride carbamate, Carbachol, Carbamylcholine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOCH2CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
182.65
Beilstein:
3917459
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carbachol

manufacturer/tradename

EDQM

mp

210 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].C[N+](C)(C)CCOC(N)=O

InChI

1S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1H

InChI key

AIXAANGOTKPUOY-UHFFFAOYSA-N

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbachol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arthur Jochems et al.
Behavioural brain research, 254, 108-115 (2013-07-03)
Medial temporal lobe (MTL) areas are crucial for memory tasks such as spatial working memory and temporal association memory, which require an active maintenance of memory for a short period of time (a few hundred milliseconds to tens of seconds).
Anna Ermund et al.
American journal of physiology. Gastrointestinal and liver physiology, 305(5), G341-G347 (2013-07-09)
Colon has been shown to have a two-layered mucus system where the inner layer is devoid of bacteria. However, a complete overview of the mouse gastrointestinal mucus system is lacking. We now characterize mucus release, thickness, growth over time, adhesive
Andrew J Hart et al.
Toxicology letters, 219(3), 262-268 (2013-04-03)
Bites by Australian black snakes (Pseudechis spp.) do not cause neurotoxicity in human envenoming. This is unusual as in vitro neurotoxicity has been reported for all Pseudechis spp. venoms. The present study aimed to identify, isolate and characterise neurotoxins from
Michael Salmon et al.
The Journal of pharmacology and experimental therapeutics, 345(2), 260-270 (2013-02-26)
Activation of muscarinic subtype 3 (M3) muscarinic cholinergic receptors (mAChRs) increases airway tone, whereas its blockade improves lung function and quality of life in patients with pulmonary diseases. The present study evaluated the pharmacological properties of a novel mAChR antagonist
Julie Svalø et al.
European journal of pharmacology, 699(1-3), 101-105 (2012-12-19)
β(3)-Adrenoceptors are major players in detrusor relaxation and have been suggested as a new putative target for the treatment of overactive bladder syndrome. We determined the effects of mirabegron (YM178), a novel β(3)-adrenoceptor agonist, on carbachol-induced tone in isolated human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service