Skip to Content
Merck
All Photos(1)

Key Documents

Y0000027

Sumatriptan for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sumatriptan succinate, 3-[2-(Dimethylamino)ethyl]-N-methyl-1H-indole-5-methanesulfonamide succinate, GR-43175

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O2S · C4H6O4
CAS Number:
Molecular Weight:
413.49
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sumatriptan

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OC(=O)CCC(O)=O.CNS(=O)(=O)Cc1ccc2[nH]cc(CCN(C)C)c2c1

InChI

1S/C14H21N3O2S.C4H6O4/c1-15-20(18,19)10-11-4-5-14-13(8-11)12(9-16-14)6-7-17(2)3;5-3(6)1-2-4(7)8/h4-5,8-9,15-16H,6-7,10H2,1-3H3;1-2H2,(H,5,6)(H,7,8)

InChI key

PORMUFZNYQJOEI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sumatriptan for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Sumatriptan succinate is a 5-HT1 serotonin receptor agonist.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihiko Tomita et al.
Journal of gastroenterology and hepatology, 28(1), 106-111 (2012-09-20)
Scintigraphy is a useful noninvasive technique for assessment of gastric motility, especially emptying, but there is little knowledge of use of the technique to assess gastric accommodation. Therefore, to clarify the usefulness of scintigraphy as a technique for assessing gastric
Benjamin W Friedman et al.
Academic emergency medicine : official journal of the Society for Academic Emergency Medicine, 19(10), 1151-1157 (2012-09-22)
Patients who use an emergency department (ED) for acute migraine headaches have higher migraine disability scores, lower socioeconomic status, and are unlikely to have used a migraine-specific medication prior to presentation to the ED. The objective was to determine if
Yasunari Sakamoto et al.
World journal of gastroenterology, 18(26), 3415-3419 (2012-07-19)
To determine the effect of oral sumatriptan on gastric emptying using a continuous ¹³C breath test (BreathID system). Ten healthy male volunteers participated in this randomized, 2-way crossover study. The subjects fasted overnight and were randomly assigned to receive a
Y Sekino et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 24(12), 1083-e564-1083-e564 (2012-08-14)
Oral sumatriptan administration has been reported to delay gastric emptying after liquid meals. The aim of this study was to determine whether delayed gastric emptying is caused by enhanced gastric accommodation, impaired antral contractions, or both using ultrasonography. Ten healthy
M J Cózar-Bernal et al.
Journal of pharmaceutical and biomedical analysis, 72, 251-260 (2012-10-16)
In this paper, a novel, precise, specific, accurate and rapid reversed-phase high performance liquid chromatographic method was developed, optimized and validated for determining sumatriptan succinate in niosomes with the best chromatographic peak resolution, reduced run time and low cost of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service