Skip to Content
Merck
All Photos(1)

Key Documents

Y0000012

Hymecromone impurity A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Resorcinol, 1,3-Benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
906905
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

3.8 (vs air)

vapor pressure

1 mmHg ( 21.1 °C)

API family

hymecromone

autoignition temp.

1126 °F

manufacturer/tradename

EDQM

bp

178 °C/16 mmHg (lit.)

mp

109-112 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1cccc(O)c1

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hymecromone impurity A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 1 Oral - STOT SE 2 Oral

Target Organs

Central nervous system,Blood, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry S Lynch et al.
Regulatory toxicology and pharmacology : RTP, 36(2), 198-210 (2002-12-04)
Resorcinol administered at high doses to rodents can disrupt thyroid hormone synthesis and can produce goitrogenic effects. These effects were not seen in a 2-year bioassay at doses of up to 520 mg/kg/day. There are species-specific differences in synthesis, binding
Resorcinol.
IARC monographs on the evaluation of carcinogenic risks to humans, 71 Pt 3, 1119-1131 (1999-09-07)
50% resorcinol peel.
P G Karam
International journal of dermatology, 32(8), 569-574 (1993-08-01)
Frank Welsch
International journal of toxicology, 27(1), 59-63 (2008-02-23)
Medical case reports published in the 20th century over the course of several decades show that resorcinol caused reversible adverse effects on the human thyroid gland (TG) manifested as hypothyroidism. Affected patients had ulcerating leg varicosities and underwent prolonged treatment
Satoshi Yamaori et al.
Chemico-biological interactions, 215, 62-68 (2014-03-29)
We previously demonstrated that cannabidiol (CBD) was a potent mechanism-based inhibitor of human cytochrome P450 1A1 (CYP1A1). However, the moiety of CBD that contributes to the potent mechanism-based inhibition of human CYP1A1 remains unknown. Thus, the effects of compounds structurally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service