Skip to Content
Merck
All Photos(1)

Key Documents

XSCRT

Sigma-Aldrich

Xscript I+DNA-Y cDNA Synthesis Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

form

liquid

Quality Level

technique(s)

PCR: suitable
qPCR: suitable

shipped in

dry ice

storage temp.

−20°C

General description

The Xscript I + DNA-Y cDNA Synthesis Kit includes a new generation of reverse transcription more suitable for optimization, buffers that further improve the efficiency of one-strand synthesis, and single-component reverse transcription primers Oligo (dT)20VN and Random hexamers allowing for increased user flexibility.

Features and Benefits

The Xscript I + DNA-Y cDNA Synthesis Kit has the following advantages:
  • Users can choose reverse transcription primers for subsequent experiments according to their needs.
  • Kit can synthesize full-length cDNA (up to 20 kb) for downstream experiments such as cloning.
  • Kit can synthesize highly uniform cDNA for qPCR quantification.
  • 5 × gDNA wiper Mix in this kit can quickly remove genomic DNA contamination at 42°C for 2 min.
  • Ensures more reliable follow-up results.
  • Simplifies qPCR primer design without the need to design primers across introns.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.

Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.

Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.

Explore high-quality raw materials for diagnostic assay development and IVD manufacturing. Leverage our expertise in molecular diagnostic testing and enzymes for molecular assays.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service