Skip to Content
Merck
All Photos(1)

Key Documents

U0020008

Ubidecarenone impurity D

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Coenzyme Q9, Q-9, Ubiquinone-45, Ubiquinone-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H82O4
CAS Number:
Molecular Weight:
795.23
Beilstein:
1900081
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

ubidecarenone

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COC1=C(OC)C(=O)C(C\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C)=C(C)C1=O

InChI

1S/C54H82O4/c1-40(2)22-14-23-41(3)24-15-25-42(4)26-16-27-43(5)28-17-29-44(6)30-18-31-45(7)32-19-33-46(8)34-20-35-47(9)36-21-37-48(10)38-39-50-49(11)51(55)53(57-12)54(58-13)52(50)56/h22,24,26,28,30,32,34,36,38H,14-21,23,25,27,29,31,33,35,37,39H2,1-13H3/b41-24+,42-26+,43-28+,44-30+,45-32+,46-34+,47-36+,48-38+

InChI key

UUGXJSBPSRROMU-WJNLUYJISA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous detection of ubiquinol-10, ubiquinone-10, and tocopherols in human plasma microsamples and macrosamples as a marker of oxidative damage in neonates and infants
Menke T, et al.
Analytical Biochemistry, 282(2), 209-217 (2000)
Leilei Li et al.
International journal of systematic and evolutionary microbiology, 64(Pt 7), 2407-2415 (2014-04-26)
Five acetic acid bacteria isolates, awK9_3, awK9_4 ( = LMG 27543), awK9_5 ( = LMG 28092), awK9_6 and awK9_9, obtained during a study of micro-organisms present in traditionally produced kefir, were grouped on the basis of their MALDI-TOF MS profile with LMG
Juquan Jiang et al.
International journal of systematic and evolutionary microbiology, 64(Pt 5), 1662-1669 (2014-02-11)
A moderately halophilic bacterium (strain NEAU-ST10-39T) was isolated from saline and alkaline soils in the oilfield of Daqing City, Heilongjiang Province, China. The strain was strictly aerobic, Gram-stain-negative, rod-shaped and motile by peritrichous flagella. Its colonies were yellow. It grew
Asif Hameed et al.
International journal of systematic and evolutionary microbiology, 64(Pt 7), 2330-2337 (2014-04-20)
Two Gram-staining-negative, aerobic, rod-shaped, non-spore-forming bacterial strains that are motile by a monopolar flagellum, designated CC-AMH-11(T) and CC-AMHZ-5, were isolated from droppings of a seashore bird off the coast of Hualien, Taiwan. The strains showed 99.7% mutual pairwise 16S rRNA
Wen-Bo Tan et al.
International journal of systematic and evolutionary microbiology, 65(Pt 1), 225-229 (2014-10-19)
A Gram-staining-negative, rod-shaped, motile and facultatively anaerobic bacterial strain, designated X2(T), was isolated from the sludge of an anaerobic, denitrifying, sulfide-removal bioreactor, and found to oxidize sulfide anaerobically with nitrate as electron acceptor. The strain grew at salinities of 0-3%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service