TDS1000
Total Dissolved Solids 1000 mg/L Calibration Standard
certified reference material
Synonym(s):
TDS 1000 mg/L Reference Solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
CofA
current certificate can be downloaded
manufacturer/tradename
RTC TDS1000-100
concentration
in H2O
application(s)
environmental
format
matrix material
storage temp.
2-30°C
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Total Dissolved Solids 1000 mg/L Calibration Standard for R&D and Analytical use only.
Features and Benefits
- Product is packed in High Density Polyethylene (HDPE) bottle
- A certificate of analysis (CoA) is included with the product
- Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and International System of units (SI)
- Gravimetrically performed dilutions
- Stable, ready to use solution with long minimum shelf life
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service