The Critical Micellar Concentration (CMC) of Ecosurf SA-9 is 22 ppm, or 0.0022 percent, at 25°C. So one would need to use a concentration of greater than 0.0022 percent. The suggestion would be to try, for example, 0.01%, 0.1%, and 1% side-by-side to see which gives the best results, and optimize from there. The optimal concentration will vary, depending on the application.
STS0007
ECOSURF™ SA-9
Non-ionic surfactant
Synonym(s):
Alcohols ethoxylated propoxylated, Seed oil alcohol ethoxylate 9 EO
About This Item
Recommended Products
grade
laboratory grade
Quality Level
description
non-ionic
form
liquid
greener alternative product characteristics
Design for Degradation
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
CMC
22 ppm (25°C)
viscosity
30.225 cSt(40 °C)
transition temp
pour point 4 °C
cloud point 57 °C (1 wt% actives aq solution)
solubility
water: freely soluble (some compositions may form gels)
density
0.9831 g/mL at 20 °C
HLB
11 - 13
greener alternative category
, Aligned
SMILES string
O(CCCCCC)CCC.O(CCCCCC)CC
InChI
1S/C9H20O.C8H18O/c1-3-5-6-7-9-10-8-4-2;1-3-5-6-7-8-9-4-2/h3-9H2,1-2H3;3-8H2,1-2H3
InChI key
QTUGCFPIWBMXQE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Superior wetting
- Excellent detergency
- Rapid dissolution & good rinseability
- Low odor
- No gel range
- Outstanding formulation & handling properties
- Readily biodegradable (OECD 301)
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
I'd like to test ecosurf sa-9 as an alternative to Triton X-100 for the lysis of virus or animal cell (RBC). Currently, we utilize the aqueous 1% Triton X-100 solution. Can you recommend the concentration range of ecosurf sa-9 for such prupose ?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service