SQC005
TCLP Metals in Soil
certified reference material
Synonym(s):
Soil matrix CRM, TCLP metals
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
CofA
current certificate can be downloaded
manufacturer/tradename
RTC SQC-005
technique(s)
AAS: suitable
ICP: suitable
matrix
soil
application(s)
environmental
format
matrix material
storage temp.
2-30°C
Related Categories
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
<ul>
<li><strong>Soil toxicity assessment for biotech development sites:</strong> TCLP and <em>in vitro</em> PBET model were used to determine the leachability (ecological risk) and oral bioaccessibility (human health risk) of soil cadmium (Chen et al., 2024).</li>
<li><strong>TCLP compliance testing for biopharmaceutical researchers: </strong>Used in leachability of heavy metals. The biomineralization driven by <em>B. pasteurii</em> on Cd and Pb in the presence of a calcium source was evaluated based on the contents of Cd and Pb using TCLP (Zhang et al., 2024).</li>
</ul>
<li><strong>Soil toxicity assessment for biotech development sites:</strong> TCLP and <em>in vitro</em> PBET model were used to determine the leachability (ecological risk) and oral bioaccessibility (human health risk) of soil cadmium (Chen et al., 2024).</li>
<li><strong>TCLP compliance testing for biopharmaceutical researchers: </strong>Used in leachability of heavy metals. The biomineralization driven by <em>B. pasteurii</em> on Cd and Pb in the presence of a calcium source was evaluated based on the contents of Cd and Pb using TCLP (Zhang et al., 2024).</li>
</ul>
Other Notes
Values of analytes vary lot to lot. Some lots may not contain all analytes.
Disclaimer
For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Skin Sens. 1
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service