Skip to Content
Merck
All Photos(1)

Key Documents

SMB00963

Sigma-Aldrich

Desethylchloroquine

research grade, ≥95% (HPLC)

Synonym(s):

7-Chloro-4-{[4-(ethylamino)-1-methylbutyl]amino}quinoline, 4-[[1-Methyl-4-((ethylamino)butyl]amino)]-7-chloroquinoline, 7-Chloro-4-(4-N-ethylamino-1-methylbutylamino)quinoline, N-Desethyl chloroquine, N-Desethylchloroquine, N4-(7-Chloro-4-quinolinyl)-N1-ethyl-1,4-pentanediamine, Monodeethylchloroquine, Monodesethylchloroqu​ine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22ClN3
CAS Number:
Molecular Weight:
291.82
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥95% (HPLC)

form

powder or crystals

technique(s)

HPLC: suitable

color

white to off-white

shipped in

wet ice

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC2=C1C(NC(CCCNCC)C)=CC=N2

InChI

1S/C16H22ClN3/c1-3-18-9-4-5-12(2)20-15-8-10-19-16-11-13(17)6-7-14(15)16/h6-8,10-12,18H,3-5,9H2,1-2H3,(H,19,20)

InChI key

MCYUUUTUAAGOOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Desethylchloroquine is a metabolite of hydroxychloroquine and chloroquine. Hydroxychloroquine and chloroquine have antimalarial properties and are used as a therapeutic for rheumatoid arthritis (RA), systemic lupus erythematosus (SLE) and other inflammatory rheumatic diseases. They have also been in the spotlight due to their activity on patients with COVID-19, the disease caused by the SARS-CoV-2 virus. Hydroxychloroquine and chloroquine interfere with lysosomal activity and autophagy, interact with membrane stability and alter signalling pathways and transcriptional activity, which can result in modulation of cytokine production. Their mode of action is still emerging and their metabolism is of great interest.

Application

Application: Metabolomics research

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaëlle Noé et al.
Therapeutic drug monitoring, 41(4), 476-482 (2019-02-27)
Hydroxychloroquine (HCQ) is approved for the treatment of systemic lupus erythematosus (SLE). Therapeutic drug monitoring of HCQ is necessary to detect nonadherence and to improve treatment efficacy in patients with SLE. Liquid chromatographic-tandem mass spectroscopy and high performance liquid chromatography
Karnrawee Kaewkhao et al.
Bioanalysis, 11(5), 333-347 (2019-03-16)
Chloroquine is an antimalarial drug used in the treatment of Plasmodium vivax malaria. Three methods to quantify chloroquine and its metabolite in blood matrices were developed and validated. Methodology & results: Different high-throughput extraction techniques were used to recover the
Yingshan Han et al.
Journal of medical virology, 91(7), 1182-1190 (2019-02-26)
Studies aimed at repurposing existing drugs revealed that some antimalarial compounds possess anti-Zika virus (anti-ZIKV) activity. Here, we further tested 14 additional antimalarial drugs and their metabolites or analogs for anti-ZIKV activity using a phenotypic screening approach. We identified four

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service