Skip to Content
Merck
All Photos(3)

Key Documents

S7920

Sigma-Aldrich

Sodium fluoride

BioXtra, ≥99%

Synonym(s):

NaF, Natrium fluoride, Sodium monoflouride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25

vapor pressure

1.4 mmHg ( 0 °C)

Quality Level

product line

BioXtra

Assay

≥99%

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter

mp

993 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.03%

cation traces

Al: ≤0.001%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.003%
K: ≤0.02%
Mg: ≤0.0005%
NH4+: ≤0.1%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium fluoride has been used:
  • as a source of fluoride ions, to analyse its impact on spermatogenesis and capacitation in vitro
  • as an additive in the extraction buffer for the isolation of nuclei, cytoplasmic and nuclear proteins
  • as a component of lysis buffer for the lysis of human ovarian adenocarcinoma OVCAR3 cells

Biochem/physiol Actions

Sodium fluoride (NaF) prevents dental caries, by catalysing the diffusion of calcium and phosphate and remineralizes the lesions. Fluoride affects spermatogenesis and capacitation process in the female reproductive tract. Fluoride induces oxidative stress, DNA damage in ovary ultimately affects the development of oocytes. Fluoride supplementation enhances bone mineral density. 18F-NaF is a radiotracer used in positron emission tomography (PET) to identify atherosclerotic lesions and other cardiac plaques.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mangiferin induces apoptosis in human ovarian adenocarcinoma OVCAR3 cells via the regulation of Notch3
Zou B, et al.
Oncology Reports, 38(3), 1431-1441 (2017)
Dental caries
Selwitz RH, et al.
Lancet, 369(9555), 51-59 (2007)
Fluoride-induced oxidative stress and apoptosis are involved in the reducing of oocytes development potential in mice
Wang HW, et al.
Chemosphere, 186, 911-918 (2017)
Effects of treatment with fluoride on bone mineral density and fracture risk-a meta-analysis
Vestergaard P, et al.
Osteoporosis International : A Journal Established as Result of Cooperation Between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA, 19(3), 257-268 (2008)
Toxicological Profile for Fluorides, Hydrogen Fluoride, and Fluorine (Update) (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service