Skip to Content
Merck
All Photos(1)

Key Documents

S0947000

Somatropin

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

somatropin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Somatropin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kuzma et al.
Bratislavske lekarske listy, 114(12), 689-695 (2013-12-18)
Growth hormone deficiency (GHD) is associated with reduced bone mineral content and increased risk of osteoporotic fractures. Reduced peak bone mass might explain the low bone mineral density (BMD) among patients with childhood onset GHD (CO-GHD) whilst the cause of
O V Bol'shova et al.
Likars'ka sprava, (5)(5), 70-75 (2014-03-13)
The objective was to study blood Zinc levels and hair Zinc content in three subsets of 40 naive patients (29 boys) with childhood onset GH deficiency (isolated total GHD, partial GHD, syndrome biological inactive GH). Our data showed that GHD
Ralf Nass et al.
The Journal of clinical endocrinology and metabolism, 99(2), 602-608 (2013-11-29)
Acyl-ghrelin is thought to have both orexigenic effects and to stimulate GH release. A possible cause of the anorexia of aging is an age-dependent decrease in circulating acyl-ghrelin levels. The purpose of the study was to compare acyl-ghrelin and GH
Bilal Battal et al.
The neuroradiology journal, 27(1), 55-62 (2014-02-28)
Brain herniations into dural venous sinuses (DVS) are rare findings recently described and their etiology and clinical significance are controversial. We describe five patients with brain herniations into the DVS or calvarium identified on MRI, and discuss their imaging findings
Małgorzata Partyka et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 36(211), 63-67 (2014-03-22)
Growth hormone (GH) is a polypeptide hormone produced by the cells of pituitary. Production of growth hormone is carried out in a pulsating manner, and the frequency and intensity of the pulses is dependent on age and gender. Growth hormone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service