Skip to Content
Merck
All Photos(1)

Key Documents

R0950000

Rifamycin S

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Dideoxy-1,4-dihydro-1,4-dioxorifamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H45NO12
CAS Number:
Molecular Weight:
695.75
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifamycin, rifampicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1C2=CC(=O)c3c4c(c(c(c3C2=O)O)C)O[C@](O\C=C\[C@@H]([C@H]([C@H]([C@@H]([C@@H]([C@@H]([C@H]([C@H](\C=C\C=C(/C1=O)\C)C)O)C)O)C)OC(=O)C)C)OC)(C4=O)C

InChI

1S/C37H45NO12/c1-16-11-10-12-17(2)36(46)38-23-15-24(40)26-27(32(23)44)31(43)21(6)34-28(26)35(45)37(8,50-34)48-14-13-25(47-9)18(3)33(49-22(7)39)20(5)30(42)19(4)29(16)41/h10-16,18-20,25,29-30,33,41-43H,1-9H3,(H,38,46)/b11-10+,14-13+,17-12-/t16-,18+,19+,20+,25-,29-,30+,33+,37-/m0/s1

InChI key

BTVYFIMKUHNOBZ-ODRIEIDWSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifamycin S EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT SE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiqin Huang et al.
Antonie van Leeuwenhoek, 95(2), 143-148 (2009-01-07)
Strain AM105 was separated from mangrove sediment in the South China Sea in this research. The morphological and genomic data showed that the strain merits description as a novel species, proposed as Micromonospora rifamycinica. From the acetate ethyl extract of
Eduardo Rubio et al.
Magnetic resonance in chemistry : MRC, 43(4), 269-282 (2005-01-28)
New spiro-piperidylrifamycin derivatives are presented. These compounds were synthesized from the reaction of 3-amino-4-iminorifamycin S and enantiomerically pure 4-piperidones, which generate diasteroisomeric rifabutin analogues with a new stereocentre at the spiranic carbon. The (1)H and (13)C NMR spectra of these
U C Banerjee
Enzyme and microbial technology, 15(12), 1037-1041 (1993-12-01)
Growing and resting cell systems of Curvularia lunata were used for the transformation of rifamycin B to rifamycin S. In the case of growing cells, rifamycin B was added at the time of inoculation and at the different phases of
K W Hunt et al.
Organic letters, 3(3), 481-484 (2001-06-29)
[figure: see text] The direct opening at the bridgehead of oxabicyclo[3.2.1]octenes employing silyl ketene acetals in 4.0-5.0 M lithium perchlorate in diethyl ether has been realized, which gives rise to highly functionalized cycloheptadienes that can be further manipulated for use
Ming Chen et al.
The Journal of organic chemistry, 78(1), 3-8 (2012-06-19)
Syntheses of the C(15)-C(27) fragments of chaxamycins A/D, rifamycin S, and the C(12)-C(24) fragment of salinisporamycin have been accomplished in 10 steps from commercially available starting materials. Three crotylboron reagents were utilized to construct the seven contiguous stereocenters in these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service