PHR2829
Adapalene Related Compound C
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
o-Adamantylanisole, Adapalene Impurity C (PhEur), 1-(2-Methoxyphenyl)-tricyclo[3.3.1.13,7]decane, 2-(Adamant-1-yl)methoxybenzene
About This Item
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to Ph. Eur. Y0001120
traceable to USP 1011732
API family
adapalene
form
solid
packaging
pkg of 50 mg
application(s)
pharmaceutical small molecule
storage temp.
2-8°C
SMILES string
O(C)c1c(cccc1)C32CC4CC(C3)CC(C2)C4
InChI
1S/C17H22O/c1-18-16-5-3-2-4-15(16)17-9-12-6-13(10-17)8-14(7-12)11-17/h2-5,12-14H,6-11H2,1H3
InChI key
HYSZKSPAPGPYFQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Other Notes
Footnote
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service