PHR2583
Memantine Related Compound D
Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 100 mg
Synonym(s):
1-Bromo-3,5-dimethyladamantane, 1-Bromo-3,5-dimethyltricyclo[3.3.1.13,7]decane
Select a Size
Select a Size
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1380546
API family
memantine
form
liquid
packaging
pkg of 100 mg
refractive index
n20/D 1.52 (lit.)
bp
201 °C (lit.)
density
1.224 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
related product
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service