Skip to Content
Merck
All Photos(1)

Key Documents

PHR1470

Supelco

Scopolamine Hydrobromide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Scopolamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HBr · 3H2O
CAS Number:
Molecular Weight:
438.31
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 199
traceable to Ph. Eur. H1500000
traceable to USP 1610001

API family

scopolamine, hyoscine

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Br.N1([C@@H]2[C@@H]3O[C@@H]3[C@H]1C[C@H](C2)OC(=O)[C@H](CO)c4ccccc4)C

InChI

1S/C17H21NO4.BrH/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10;/h2-6,11-16,19H,7-9H2,1H3;1H/t11-,12-,13-,14+,15-,16+;/m1./s1

InChI key

WTGQALLALWYDJH-MOUKNHLCSA-N

Gene Information

human ... CHRM1(1128)

General description

Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards
Scopolamine Hydrobromide is the hydrobromide salt of scopolamine, naturally found in belladonna plant. It belongs to the group of cycloplegic drugs, which produces cycloplegia. Its mode of action involves inhibiting the action of acetylcholine at muscarinic receptors in the ciliary muscle.

Application

Scopolamine Hydrobromide may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB7821 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dictionary of Optometry and Visual Science, 590(2), 341-353 (2014)
Topical Drug Bioavailability, Bioequivalence, and Penetration, 590(2), 341-353 (1993)
Comparison of isotachophoresis, capillary zone electrophoresis and high-performance liquid chromatography for the determination of salbutamol, terbutaline sulphate and fenoterol hydrobromide in pharmaceutical dosage forms
Ackermans TM, et al.
Journal of Chromatography A, 590(2), 341-353 (1992)
LC determination of atropine sulfate and scopolamine hydrobromide in pharmaceuticals.
Ceyhan T, et al.
Journal of Pharmaceutical and Biomedical Analysis, 25(3-4), 399-406 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service