Skip to Content
Merck
All Photos(1)

Key Documents

PHR1446

Supelco

Ramipril

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Tritace, [2S,3aS,6aS]-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]octahydrocyclopenta[b]pyrrole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32N2O5
CAS Number:
Molecular Weight:
416.51
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 751
traceable to Ph. Eur. R0145000
traceable to USP 1598303

API family

ramipril

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(N1[C@](CCC2)([H])[C@]2([H])C[C@H]1C(O)=O)[C@H](C)N[C@H](C(OCC)=O)CCC3=CC=CC=C3

InChI

1S/C23H32N2O5/c1-3-30-23(29)18(13-12-16-8-5-4-6-9-16)24-15(2)21(26)25-19-11-7-10-17(19)14-20(25)22(27)28/h4-6,8-9,15,17-20,24H,3,7,10-14H2,1-2H3,(H,27,28)/t15-,17-,18-,19-,20-/m0/s1

InChI key

HDACQVRGBOVJII-JBDAPHQKSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Ramipril is an angiotensin-converting-enzyme (ACE) inhibitor, which acts on the renin angiotensin aldosterone system. It is widely used in reducing hypertension by the suppression of the effects of angiotensin II at its receptor, thereby blocking the renin-angiotensin system.

Application

Ramipril may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique and high-performance liquid chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Angiotensin converting enzyme (ACE) inhibitor.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3757 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Spectrophotometric method for the determination of ramipril in solid dosage forms
Afleroho EO, et al.
Tropical Journal of Pharmaceutical Research, 11(2), 275-279 (2012)
A validated HPLC method for analysis of atorvastatin calcium, ramipril and aspirin as the bulk drug and in combined capsule dosage Forms
Patole MS, et al.
International Journal of Pharmaceutical Sciences Review and Research, 4(3), 40-45 (2010)
Simultaneous determination of ramipril, hydrochlorothizide and telmisartan by spectrophotometry
Bankey S, et al.
International Journal of ChemTech Research, 1(2), 183-188 (2009)
Development and validation of simultaneous HPLC method for estimation of telmisartan and ramipril in pharmaceutical formulations
Jawla S, et al.
International Journal of PharmTech Research, 2(2), 1625-1633 (2010)
Roberto Lozano et al.
International journal of clinical pharmacology and therapeutics, 53(3), 206-210 (2015-02-12)
Our aim was to determinate the acenocoumarol dose requirement in highly sensitive geriatric patients, based on a minimum of genotype (VKORC1 and CYP2C9) data. We used a Gaussian kernel density estimation test to identify patients highly sensitive to the drug

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service