Skip to Content
Merck

PHR1361

Supelco

Calcium Ascorbate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Calcium L-ascorbate dihydrate, L-Ascorbic acid calcium salt dihydrate, Vitamin C calcium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14CaO12 · 2H2O
CAS Number:
Molecular Weight:
426.34
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1086356

API family

calcium ascorbate

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

166 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB3501224SAB3501205SAB3501203
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

biological source

goat

biological source

goat

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

UniProt accession no.

Q692M2

UniProt accession no.

Q692M2

UniProt accession no.

Q692M2

UniProt accession no.

Q710U6

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Calcium Ascorbate is a non-acidic buffered form of ascorbate, which is found to be safer for mouth and sensitive stomach. It can also be directly applied to gums to heal infections.[1]

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3786 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

How to Stop Colds, Allergies & More (2012)
How to Stop Colds, Allergies & More (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service