Skip to Content
Merck
All Photos(1)

Key Documents

PHR1296

Supelco

Bifonazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(p,α-Diphenylbenzyl)imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. B1110000

API family

bifonazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

c1ccc(cc1)C(c2ccc(cc2)-c3ccccc3)n4ccnc4

InChI

1S/C22H18N2/c1-3-7-18(8-4-1)19-11-13-21(14-12-19)22(24-16-15-23-17-24)20-9-5-2-6-10-20/h1-17,22H

InChI key

OCAPBUJLXMYKEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Bifonazole is a substituted imidazole analog with therapeutic potential in treating invasive mucosal infections. It exhibits antifungal activity by suppressing the proliferation of dermatophytes, yeasts and fungi affecting the skin and nails. Its mode of action involves blocking the fungal ergosterol biosynthetic pathway and additional inhibition of terpenoid biosynthesis.

Application

Bifonazole may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Bifonazole is an imidazole-based anti-fungal agent with broad spectrum activity against many fungi, molds, yeast and some Gram-positive bacteria. Bifonazole inhibits ergosterol biosynthetic protein 28 and Cytochrome P450 2B4.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC3256 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validated spectrophotometric methods for the determination of bifonazole in pharmaceuticals by charge transfer complexation
Ismail SBN, et al.
Journal of the Association of Arab Universities for Basic and Applied Sciences, 19(1), 8-14 (2016)
Determination of bifonazole in creams containing methyl-and propyl p-hydroxybenzoate by derivative spectrophotometric method
Popovic G, et al.
Journal of Pharmaceutical and Biomedical Analysis, 33(1), 131-136 (2003)
Betty Wächtler et al.
Antimicrobial agents and chemotherapy, 55(9), 4436-4439 (2011-07-13)
Clotrimazole and bifonazole are highly effective antifungal agents against mucosal Candida albicans infections. Here we examined the effects of low levels of clotrimazole and bifonazole on the ability of C. albicans to adhere, invade, and damage vaginal epithelial cells. Although
Yonghong Zhao et al.
The Journal of biological chemistry, 281(9), 5973-5981 (2005-12-24)
To better understand ligand-induced structural transitions in cytochrome P450 2B4, protein-ligand interactions were investigated using a bulky inhibitor. Bifonazole, a broad spectrum antifungal agent, inhibits monooxygenase activity and induces a type II binding spectrum in 2B4dH(H226Y), a modified enzyme previously
Salomé El Hage et al.
Archiv der Pharmazie, 344(6), 402-410 (2011-03-25)
Two series of chlorinated benzhydryl imidazole and triazole derivatives were synthesized and tested in vitro against representative strains of potent pathogenic bacteria (Staphylococcus aureus CIP 4.83, Escherichia hirae CIP 5855, Pseudomonas aeruginosa CIP 82118, Escherichia coli CIP 53126) and fungi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service