Skip to Content
Merck
All Photos(1)

Key Documents

PHR1160

Supelco

Fluconazole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 1174
traceable to Ph. Eur. Y0000557
traceable to USP 1271700

API family

fluconazole

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

FC1=CC(F)=C(C(CN2N=CN=C2)(O)CN3N=CN=C3)C=C1

InChI

1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2

InChI key

RFHAOTPXVQNOHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Fluconazole is an antifungal agent with broad-spectrum antimicrobial activity. It is employed for the remediation of oral, esophagal and vaginal candidiasis and other serious systemic candidal infections. The drug is also effective against meningitis caused by Cryptococcus neoformans.

Application

Fluconazole may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography and spectrophotometry techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Fluconazole is an antifungal agent. It is highly selective inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Fluconazole is a potent inhibitor of CYP2C9. Fluconazole interferes with fungal ergosterol synthesis and downregulates the metallothionein gene.
Fluconazole is an antifungal agent; highly selctive inhibitor of fungal cytochrome P-450 sterol C-14 α-demethyllation. Potent inhibitor of CYP2C9. Interferes with fungal ergosterol synthesis; downregulates metallothionein gene.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Values of analytes vary lot to lot.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0255 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Lact. - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

QUANTITATIVE DETERMINATION OF FLUCONAZOLE IN CAPSULES AND IV SOLUTIONS BY UV SPECTROPHOTOMETRIC METHODS
Go?er NG, et al.
Analytical Letters, 34(12), 2089-2098 (2001)
Joseph H Skalski et al.
PLoS pathogens, 14(9), e1007260-e1007260 (2018-09-21)
The gastrointestinal microbiota influences immune function throughout the body. The gut-lung axis refers to the concept that alterations of gut commensal microorganisms can have a distant effect on immune function in the lung. Overgrowth of intestinal Candida albicans has been
QUANTITATIVE DETERMINATION OF FLUCONAZOLE IN SYRUPS BY FIRST ORDER DERIVATIVE SPECTROPHOTOMETRY
Aboul-Enein HY, et al.
Analytical Letters, 35(7), 1193-1204 (2002)
Evaluation of the bioequivalence of capsules containing 150 mg of fluconazole
Porta V, et al.
International Journal of Pharmaceutics, 288(1), 81-86 (2005)
A-C L Meyer et al.
Tropical medicine & international health : TM & IH, 18(4), 495-503 (2013-02-02)
To test the hypothesis that a screening and treatment intervention for early cryptococcal infection would improve survival among HIV-infected individuals with low CD4 cell counts. Newly enrolled patients at Family AIDS Care and Education Services (FACES) in Kenya with CD4 ≤ 100

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service