Skip to Content
Merck
All Photos(1)

Key Documents

PHR1108

Supelco

L-Histidine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

(S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O2
CAS Number:
Molecular Weight:
155.15
Beilstein:
84088
EC Number:
MDL number:
UNSPSC Code:
41116107
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. H0750000
traceable to USP 1308505

API family

histidine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

282 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

N[C@@H](Cc1c[nH]cn1)C(O)=O

InChI

1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1

InChI key

HNDVDQJCIGZPNO-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. L-Histidine is an essential amino acid, widely present at the active sites of enzymes. It binds to metal ions and may aid in the transport of copper. It also plays an important role for the binding of heme groups, macromolecules and phosphate groups in the binding sites of proteins.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Precursor of histamine by action of histidine decarboxylase.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7159 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shweta Singh et al.
Microbial cell factories, 7, 28-28 (2008-10-14)
The large-scale production of G-protein coupled receptors (GPCRs) for functional and structural studies remains a challenge. Recent successes have been made in the expression of a range of GPCRs using Pichia pastoris as an expression host. P. pastoris has a
Pawel Swietach et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(22), E2064-E2073 (2013-05-17)
Ca(2+) signaling regulates cell function. This is subject to modulation by H(+) ions that are universal end-products of metabolism. Due to slow diffusion and common buffers, changes in cytoplasmic [Ca(2+)] ([Ca(2+)]i) or [H(+)] ([H(+)]i) can become compartmentalized, leading potentially to
H M Golder et al.
Journal of dairy science, 97(8), 5131-5150 (2014-06-03)
Ruminal bacterial community composition (BCC) and its associations with ruminal fermentation measures were studied in dairy heifers challenged with combinations of grain, fructose, and histidine in a partial factorial study. Holstein-Friesian heifers (n=30) were randomly allocated to 5 triticale grain-based
Giorgio Zoppellaro et al.
Biopolymers, 91(12), 1064-1082 (2009-06-19)
Six-coordinated heme groups are involved in a large variety of electron transfer reactions because of their ability to exist in both the ferrous (Fe(2+)) and ferric (Fe(3+)) state without any large differences in structure. Our studies on hemes coordinated by
Tomas F M Kuijpers et al.
The FEBS journal, 280(23), 6184-6195 (2013-10-12)
Sulfite salts are widely used as antibrowning agents in food processing. Nevertheless, the exact mechanism by which sulfite prevents enzymatic browning has remained unknown. Here, we show that sodium hydrogen sulfite (NaHSO3) irreversibly blocks the active site of tyrosinase from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service