Skip to Content
Merck
All Photos(1)

Key Documents

PHR1089

Supelco

Cimetidine hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N6S · HCl
CAS Number:
Molecular Weight:
288.80
Beilstein:
5668216
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C2175500
traceable to USP 1134073

API family

cimetidine

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.CN\C(NC#N)=N/CCSCc1nc[nH]c1C

InChI

1S/C10H16N6S.ClH/c1-8-9(16-7-15-8)5-17-4-3-13-10(12-2)14-6-11;/h7H,3-5H2,1-2H3,(H,15,16)(H2,12,13,14);1H

InChI key

QJHCNBWLPSXHBL-UHFFFAOYSA-N

Gene Information

human ... HRH2(3274)

Looking for similar products? Visit Product Comparison Guide

General description

Cimetidine is a histamine H2-receptor antagonist, widely used to inhibit gastric acid secretion. It finds application in the treatment of gastric and duodenal ulcers.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Cimetidine may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical preparations using gas chromatography technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7168 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

GC Determination of famotidine, ranitidine, cimetidine, and metformin in pharmaceutical preparations and serum using methylglyoxal as derivatizing reagent
Majidano AS, et al.
Chromatographia, 75(21-22), 1311-1317 (2012)
Duo-Yu Wu et al.
Asian Pacific journal of tropical medicine, 5(10), 831-833 (2012-10-10)
To evaluate the clinical effects of psychological interventions and drug therapy against peptic ulcer. 96 patients with peptic ulcer were divided into control group with Tagamet 800 mg per evening p.o. and trial group with psychological intervention on the basis
Yisheng Zheng et al.
Molecular immunology, 54(1), 74-83 (2012-12-12)
Cimetidine, a histamine type-2 receptor antagonist, is known to inhibit the growth of several tumors in human and animals, however the mechanism of action underlying this effect remains largely unknown. Here, in the mice model of 3LL lung tumor, cimetidine
Toru Arino et al.
European journal of pharmacology, 693(1-3), 72-79 (2012-09-13)
The antiprotozoal agent pentamidine, used for the treatment of Pneumocystis jirovecii pneumonia (PCP), is known to cause abnormalities in blood glucose homeostasis, such as hypoglycemia and hyperglycemia. Pentamidine has been reported to be a substrate of organic cation transporter 1
Renata Longhini et al.
Journal of anatomy, 222(2), 239-247 (2012-12-04)
It has been demonstrated that histamine interferes with the recruitment, formation and activity of osteoclasts via H(1)- and H(2)-receptors. Cimetidine is a H(2)-receptor antagonist used for treatment of gastric ulcers that seems to prevent bone resorption. In this study, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service