All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H25NO5
CAS Number:
Molecular Weight:
299.36
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥98.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
−20°C
InChI
1S/C15H25NO5/c1-9(2)15(20,10(3)17)14(19)21-8-11-4-6-16-7-5-12(18)13(11)16/h4,9-10,12-13,17-18,20H,5-8H2,1-3H3/t10-,12-,13-,15+/m1/s1
InChI key
SFVVQRJOGUKCEG-OPQSFPLASA-N
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG.
Check out our entire range of pyrrolizidine alkaloid standards
Check out our entire range of pyrrolizidine alkaloid standards
Analysis Note
For the analysis of the phytotoxins. Pyrrolizidin alkaloid (PA)
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service