Skip to Content
Merck
All Photos(1)

Key Documents

P3350000

Proline

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Proline, (S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

proline

manufacturer/tradename

EDQM

mp

228 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Proline EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 6

1 of 6

Jiahui Jin et al.
Stem cell research & therapy, 12(1), 327-327 (2021-06-07)
Systemic sclerosis (SSc) is a disease that features severe fibrosis of the skin and lacks effective therapy. Bone marrow mesenchymal stem cell (BMSC)-derived extracellular vesicles (EVs) are potential stem cell-based tools for the treatment of SSc. BMSCs were isolated from
Antoinette Bugyei-Twum et al.
Cardiovascular diabetology, 13, 89-89 (2014-06-03)
Despite advances in the treatment of heart failure, mortality remains high, particularly in individuals with diabetes. Activated transforming growth factor beta (TGF-β) contributes to the pathogenesis of the fibrotic interstitium observed in diabetic cardiomyopathy. We hypothesized that high glucose enhances
Mattias Mandorfer et al.
AIDS patient care and STDs, 28(5), 221-227 (2014-05-07)
The aim of this study was to assess the therapeutic potential of telaprevir (TPV)/boceprevir (BOC)-based triple-therapy in a complete cohort of HIV/chronic hepatitis C co-infected patients (HIV/HCV). Moreover, a case series of four HIV/HCV genotype (HCV-GT)1 patients with rapid virologic
Marco Scocchi et al.
Cellular and molecular life sciences : CMLS, 68(13), 2317-2330 (2011-05-20)
Proline-rich antimicrobial peptides are a group of cationic host defense peptides of vertebrates and invertebrates characterized by a high content of proline residues, often associated with arginine residues in repeated motifs. Those isolated from some mammalian and insect species, although
Frederic Bringaud et al.
Frontiers in bioscience (Landmark edition), 17, 349-374 (2011-12-29)
Trypanosomatids are a large family of unicellular eukaryotes, many of which are parasites in higher eukaryotes including man. Much of our understanding of metabolism in these organisms has been gained form the study of the human infective representatives (Trypanosoma brucei

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service