Skip to Content
Merck
All Photos(1)

Key Documents

P0800000

Phenazone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Antipyrine, 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Beilstein:
157775
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

phenazone

manufacturer/tradename

EDQM

mp

109-111 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1N(C(=O)C=C1C)c2ccccc2

InChI

1S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

VEQOALNAAJBPNY-UHFFFAOYSA-N

Gene Information

human ... PTGS1(5742)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Phenazone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gebrekrustos Mekonnen et al.
PLoS neglected tropical diseases, 12(9), e0006790-e0006790 (2018-09-13)
Trypanosoma evansi is mechanically transmitted by biting flies and affects camels, equines, and other domestic and wild animals in which it causes a disease called surra. At least two types of Trypanosoma evansi circulate in Ethiopia: type A, which is
Elena Lendoiro et al.
Drug testing and analysis, 9(1), 96-105 (2016-02-26)
Amphetamine-type-stimulants (ATS) are the second most commonly used group of illicit drugs worldwide. However, in the last few years, new psychoactive substances (NPS) with stimulant effects have appeared on the illegal market, which are not detected with traditional analytical methods.
Ei Mon Phyo Lwin et al.
Analytical and bioanalytical chemistry, 409(26), 6141-6148 (2017-08-27)
A first of its kind, simple, rapid, and sensitive liquid chromatography mass spectrometry (LC-MS/MS) method was developed and validated for quantification of perindopril and perindoprilat in both human plasma and breast milk. The analytes and internal standards (phenazone and acetyl
Lena Stütz et al.
Journal of chromatography. A, 1624, 461239-461239 (2020-06-17)
Effect-directed analysis (EDA) is increasingly used in environmental monitoring to detect and identify key toxicants. High-performance thin-layer chromatography (HPTLC) has proven to be a very suitable fractionation technique for this purpose. However, HPTLC is limited in its separation efficiency. Thus
Daniela Oberleitner et al.
Chemosphere, 250, 126255-126255 (2020-02-25)
Riverbank filtration (RBF) is a reliable water purification technique that has proven to be suitable for the removal of organic micropollutants. Its removal efficiency and dependency on a variety of factors such as redox conditions, temperatures, geology, travel times, level

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service