Skip to Content
Merck
All Photos(1)

Key Documents

O0250000

Oxprenolol hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(RS)-1-(2-Allyloxyphenoxy)- 3-isopropylamino-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO3 · HCl
CAS Number:
Molecular Weight:
301.81
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxprenolol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].[N+H2](C(C)C)CC(O)COc1c(cccc1)OCC=C

InChI

1S/C15H23NO3.ClH/c1-4-9-18-14-7-5-6-8-15(14)19-11-13(17)10-16-12(2)3;/h4-8,12-13,16-17H,1,9-11H2,2-3H3;1H

InChI key

COAJXCLTPGGDAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxprenolol hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P R Bontchev et al.
Journal of inorganic biochemistry, 83(1), 25-30 (2001-02-24)
The complexation between copper(II) and the antihypertensive drug oxprenolol (HOxp) was studied both in methanol and slightly alkaline aqueous media at Cu:HOxp molar ratio from 1:1 to 1:10. Copper(lI) forms two types of complexes-a mononuclear violet one, CuOxp2, with bidentately
Antonella De Rossi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 839(1-2), 6-11 (2005-12-21)
High sensitivity analysis of oxprenolol in spiked human urine has been performed by capillary zone electrophoresis (CZE) in ammonium formate buffer pH 2.5 using an uncoated capillary with 1cm length C18 on-capillary preconcentrator at the inlet side. The preconcentrator was
M A Martínez-Gómez et al.
Journal of pharmaceutical and biomedical analysis, 39(1-2), 76-81 (2005-06-02)
The intrinsic characteristics of capillary electrophoresis have made this technique a powerful tool in the chiral separation field. The present paper deals with the enantiomeric separation of oxprenolol enantiomers by affinity electrokinetic chromatography-partial filling technique using human serum albumin (HSA)
K Görlitzer et al.
Die Pharmazie, 59(10), 763-769 (2004-11-17)
The N-allyl derivatives 2 formed from the beta-blockers 1, were acetylated to give the esters 5. The 1,5-benzoxazacycloundecine 6a and the 1,10,5-benzodioxazacyclododecine 7a were isolated by ring closing metathesis (RCM) of 5a and 5b, respectively, using Grubbs catalyst in the
K Görlitzer et al.
Die Pharmazie, 59(9), 678-682 (2004-10-23)
Alprenolol (1a) reacts with an excess of bromine to yield the tribromo derivative 3a by addition and monosubstitution, while applying oxprenolol (1b) the disubstituted tetrabromo derivative 2b is obtained. The N-dealkylated substance 3c was isolated as a by-product. Heating the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service