NIST8393
Human DNA for Whole-Genome Variant Assessment (Son of Chinese Ancestry) (HG-005)
NIST®SRM®
Synonym(s):
Human DNA for Variant Calling (Han Chinese) (HG-005)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
form
liquid
packaging
pkg of 10 × 0.05 g (10 vials)
pkg of 10 μg (1 vial)
manufacturer/tradename
NIST®
application(s)
genomic analysis
storage temp.
−20°C
Related Categories
General description
Human DNA for Whole-Genome Variant Assessment Reference Material (RM) is intended for validation, optimization, and process evaluation purposes. It consists of a male whole human genome sample of East Asian (Chinese) ancestry from the Personal Genome Project (ID hu91BD69). A unit of RM 8393 consists of a vial containing human genomic DNA extracted from a single large growth of human lymphoblastoid cell line GM24631 (labeled as HG-005) from the Coriell Institute for Medical Research (Camden, NJ).
SRM 8393_SDS
SRM 8393_Cert
SRM 8393_SDS
SRM 8393_Cert
Application
Human DNA for Whole-Genome Variant Assessment Reference Material (RM) is intended for assessing the performance of human genome sequencing variant calling by obtaining estimates of true positives, false positives, and false negatives. Sequencing applications could include:
- whole genome sequencing
- whole exome sequencing
- targeted sequencing such as gene panels
Features and Benefits
- This reference material contains isolated DNA rather than live cells and is intended for research use.
- An extensive report of the investigation is available through NIST.
- Information values are provided for single nucleotide variations (SNVs), small insertions and deletions (indels), and homozygous reference genotypes.
Other Notes
- RM 8393 is stored at –20 °C at NIST but will be shipped in freezer packs and may not arrive frozen.
- Details on expiration, storage, safety, usage, and source are provided in the NIST certificate.
- Information on biomaterials, disposal, and transport is available in the SDS.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Megan H Cleveland et al.
The Journal of molecular diagnostics : JMD, 20(5), 583-590 (2018-07-01)
The National Institute of Standards and Technology has developed reference materials for five human genomes. DNA aliquots are available for purchase, and the data, analyses, and high-confidence small variant and homozygous reference calls are freely available on the web. These
Bennett O V Shum et al.
The Journal of molecular diagnostics : JMD, 19(4), 602-612 (2017-05-16)
The sensitivity and specificity of next-generation sequencing laboratory developed tests (LDTs) are typically determined by an analyte-specific approach. Analyte-specific validations use disease-specific controls to assess an LDT's ability to detect known pathogenic variants. Alternatively, a methods-based approach can be used
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service