Skip to Content
Merck
All Photos(1)

Key Documents

NIST5M

Cast Iron

NIST® SRM® 5M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material

Quality Level

packaging

pkg of 150 g

manufacturer/tradename

NIST®

format

matrix material

storage temp.

15-25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA003494SAB4501741SAB4502170
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

product line

-

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

buffered aqueous solution

form

buffered aqueous solution

Gene Information

human ... LTK(4058)

Gene Information

human ... LCK(3932)

Gene Information

human ... LCK(3932)

Gene Information

human ... MLTK(51776)

General description

This Standard Reference Material (SRM) is intended primarily for use in chemical methods of analysis. One unit of NIST5M consists of 150g of material in the form of chips sized between 0.50mm and 1.40mm sieve openings (35mesh and 14 mesh, respectively).

SRM 5M_cert

SRM 5M_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Carbon (C), Chromium (Cr), Copper (Cu), Manganese (Mn), Molybdenum (Mo), Nickel (Ni), Nitrogen (N), Phosphorus (P), Silicon (Si), Sulfur (S), Titanium (Ti), Vanadium (V)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service