NIST39J
Benzoic acid
NIST® SRM® 39j, calorimetric standard
About This Item
Recommended Products
grade
certified reference material
Quality Level
vapor density
4.21 (vs air)
vapor pressure
10 mmHg ( 132 °C)
autoignition temp.
1061 °F
packaging
pkg of 30 g
manufacturer/tradename
NIST®
bp
249 °C (lit.)
mp
121-125 °C (lit.)
format
neat
SMILES string
OC(=O)c1ccccc1
InChI
1S/C7H6O2/c8-7(9)6-4-2-1-3-5-6/h1-5H,(H,8,9)
InChI key
WPYMKLBDIGXBTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Each unit of SRM 39J consists of 30 g of crystalline material.
SRM 39J _SDS
SRM 39J _CoA
Application
Features and Benefits
- SRM 39J is available with a certificate documenting NIST-certified purity of 0.999996 mol/mol, as measured by coulometer mass spectrometer.
- The NIST certificate is provided with uncertainty calculation, handling, storage, and use instructions.
Other Notes
Benzoic acid (C7H6O2)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation
Target Organs
Lungs
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
249.8 °F - closed cup
Flash Point(C)
121 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service