NIST3667
Creatinine in frozen human urine
NIST® SRM® 3667
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
liquid
packaging
pkg of 1 x 10 mL
manufacturer/tradename
NIST®
technique(s)
LC/MS: suitable
application(s)
clinical testing
format
matrix material
storage temp.
−70°C
1 of 4
This Item | M8250 | M2933 | 69892 |
---|---|---|---|
clinical research | agriculture | agriculture | agriculture |
powder | crystalline powder | crystalline powder | powder or crystals |
1.23 g/cm3 at 20—25 °C, 1.27 g/cm3 at 20—25 °C | - | - | 10.66 g/mL |
dry at room temperature | dry at room temperature | dry at room temperature | - |
2.5-4 (25 °C, 97.6 g/L) | - | - | - |
General description
SRM 3667 _Cert
SRM 3667 _SDS
Application
Preparation Note
Other Notes
- SRM 3667 is intended for research use.
- Details on expiration, storage, safety, usage, and source are provided in the NIST certificate
- Information on biomaterials, disposal, and transport is available in the SDS
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service