NIST3165
Vanadium (V) standard solution
NIST® SRM® 3165
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 5 x 10 mL
manufacturer/tradename
NIST®
format
single component solution
1 of 4
This Item | NIST3167A | NIST3169 | NIST3137 |
---|---|---|---|
NIST® | NIST® | NIST® | NIST® |
single component solution | single component solution | single component solution | single component solution |
pkg of 5 x 10 mL | pkg of 5 x 10 mL | pkg of 50 mL | pkg of 50 mL |
100 | 100 | 100 | 100 |
General description
SRM 3165_cert
SRM 3165_SDS
Application
Features and Benefits
- Available with a certificate documenting NIST-certified mass fraction value of vanadium obtained through two methods:
- Gravimetric preparation using high-purity vanadium metal.
- Inductively coupled plasma optical emission spectrometry (ICP-OES).
- The certified value is metrologically traceable to the derived SI unit for mass fraction, expressed as milligrams pergram.
- The NIST certificate is provided with expiration certificate, storage, handling,use and maintenance instructions.
Other Notes
Vanadium (V)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
WGK
WGK 2
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service