NIST2723B
Sulfur in diesel fuel oil
NIST® SRM® 2723b, (nominal mass fraction 10mg/kg)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 100 mL
manufacturer/tradename
NIST®
application(s)
petroleum
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Certified for the analytes listed below.
Sulfur (S)
See certificate for values and more details at nist.gov/SRM
Sulfur (S)
See certificate for values and more details at nist.gov/SRM
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
141.8 °F
Flash Point(C)
61 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service