NIST1976C
Instrument Response Standard for X-Ray Powder Diffranction
pkg of 1 disc
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
packaging
pkg of 1 disc
manufacturer/tradename
NIST®
SRM® 1976c
technique(s)
diffraction/scattering: suitable
General description
This Standard Reference Material (SRM) consists of a sintered alumina disc intended for use in calibration of X-ray powder diffraction equipment with respect to line position and intensity as a function of 2θ angle. The solid form of the SRM eliminates any variability in intensity measurements introduced by the sample loading procedure. A unit of SRM 1976b consists of a sintered alumina disc approximately 25.6 mm in diameter by 2.2 mm in thickness. For more information, please refer to the SDS and COA.
SRM 1976C_cert
SRM 1976C_SDS
SRM 1976C_cert
SRM 1976C_SDS
Other Notes
Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service