NIST1568C
Rice flour

NIST®SRM® 1568c
Synonym(s):
Rice powder
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
packaging
pkg of 50 g
manufacturer/tradename
NIST®
format
matrix material
General description
Other Notes
Certified for the analytes listed below. See certificate for values and more details
Phosphorous
Trace Elements: Aluminum (Al), Arsenic (As), Bromine (Br), Cadmium (Cd), Calcium (Ca), Chlorine (Cl2), Cobalt (Co), Copper (Cu), Dimethylarsinic acid, Inorganic arsenic (iAs), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Monomethylarsonic acid (MMA), Potassium (K), Rubidium (Rb), Selenium (Se), Sodium (Na), Sulfur (S), Tin (Sn), Zinc (Zn)
Matrix Group: Cereals, Bread, Rice and Maize
Phosphorous
Trace Elements: Aluminum (Al), Arsenic (As), Bromine (Br), Cadmium (Cd), Calcium (Ca), Chlorine (Cl2), Cobalt (Co), Copper (Cu), Dimethylarsinic acid, Inorganic arsenic (iAs), Iron (Fe), Lead (Pb), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Monomethylarsonic acid (MMA), Potassium (K), Rubidium (Rb), Selenium (Se), Sodium (Na), Sulfur (S), Tin (Sn), Zinc (Zn)
Matrix Group: Cereals, Bread, Rice and Maize
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service