Skip to Content
Merck
All Photos(1)

Key Documents

N35

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 55.36 mPa.s (25 °C)

Synonym(s):

UKAS ISO/IEC17025 Certified Viscosity Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

55.36 mPa.s(25 °C)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation. Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage.

Viscosity standards are specifically designed, tested and certified for the calibration, verification and certification of glass capillary viscometers and other viscosity measuring equipment where the precise control of temperature is required.

Application

General purpose viscosity standard; UKAS ISO/IEC17025 and ISO 17034 certified has been used to validate the high-throughput viscosity measurement of inkjet formulations used in three-dimensional inkjet printing using a one-stop liquid handling station.[1]

Features and Benefits

  • Kinematic & dynamic viscosity given at all temperatures.
  • Standard Pack Size 500 ml, available also in 5 L.
  • Fully traceable to national standards.
  • Ensures full compliance to ASTM & IP test method protocol.
  • Tamper-evident security packaging.
  • Shelf life of 2 years.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-throughput characterization of fluid properties to predict droplet ejection for three-dimensional inkjet printing formulations
Zhou Z, et al.
Additive Manufacturing, 29, 100792-100792 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service