Skip to Content
Merck
All Photos(1)

Key Documents

N1250000

Norgestrel

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-Norgestrel, (17α)-(±)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

norgestrel

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C21H28O2/c1-3-20-11-9-17-16-8-6-15(22)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1

InChI key

WWYNJERNGUHSAO-XUDSTZEESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Norgestrel EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ondrej Mestak et al.
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology, 28(10), 764-766 (2012-03-10)
We report the case of a female who had suffered from progressive lymphatic malformation in the orbito-temporal region since childhood. Many surgical interventions were performed, including radical excision and shunt drainage. Despite aggressive surgical treatment, recurrence was observed after every
Nozomi Donoyama et al.
Acupuncture in medicine : journal of the British Medical Acupuncture Society, 29(4), 304-306 (2011-10-22)
A 26-year-old top female athlete (height 157 cm), who took part in competitive sport similar to middle- and long-distance running, became amenorrhoeic in February 2009. In late May 2009, athletic amenorrhoea was diagnosed and a norgestrel-ethinyl estradiol combination (norgestrel 0.5
Kai I Cheang et al.
Fertility and sterility, 96(2), 353-359 (2011-06-17)
To evaluate the effects of a commonly used combined hormonal oral contraceptive (OC) on carbohydrate metabolism in obese as compared with lean women. 6-month prospective study. Clinical research center at an academic medical center. Premenopausal nondiabetic women with body mass
Lawrence Aderemi Olatunji et al.
Clinical hemorheology and microcirculation, 54(1), 23-31 (2012-06-01)
Oral contraceptive (OC) use and increased fructose feeding have been associated with altered cardiometabolic effects. The effect of increased dietary fructose during OC use on cardiometabolic parameters is unknown. We investigated the effects of a high-fructose diet on body weight
Jeffrey T Jensen et al.
The European journal of contraception & reproductive health care : the official journal of the European Society of Contraception, 18(4), 274-283 (2013-05-04)
To determine the effect of oestradiol valerate/dienogest (E2V/DNG) versus ethinylestradiol/norgestimate (EE/NGM) on hormone-withdrawal associated symptoms (HWAS) in otherwise healthy women who had experienced at least one of these symptoms when using 21/7-day combined oral contraceptives (COCs). This phase III, parallel-group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service