Skip to Content
Merck
All Photos(1)

Key Documents

M9784

Supelco

Mersalyl acid

analytical standard

Synonym(s):

2-(3-Hydroxymercurio-2-methoxypropylcarbamoyl)phenoxyacetic acid, 2-[N-(3-Hydroxymercuri-2-methoxypropyl)carbamoyl]phenoxyacetic acid, Salyrganic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOHgCH2CH(OCH3)CH2NHCOC6H4OCH2CO2H
CAS Number:
Molecular Weight:
483.87
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85% (TLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

192-193 °C (dec.) (lit.)

solubility

NH4OH: soluble, clear to hazy

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

COC(CNC(=O)c1ccccc1OCC(O)=O)C[Hg]O

InChI

1S/C13H16NO5.Hg.H2O/c1-9(18-2)7-14-13(17)10-5-3-4-6-11(10)19-8-12(15)16;;/h3-6,9H,1,7-8H2,2H3,(H,14,17)(H,15,16);;1H2/q;+1;/p-1

InChI key

HQRSUIDICNOLPX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Moog et al.
Scientific reports, 10(1), 1167-1167 (2020-01-26)
The triose phosphate transporter (TPT) is one of the prerequisites to exchange metabolites between the cytosol and plastids. In this study, we demonstrated that the four plastid TPT homologues in the non-photosynthetic diatom Nitzschia sp. NIES-3581 were highly likely integrated
G Moreno et al.
Archives of biochemistry and biophysics, 386(2), 243-250 (2001-05-23)
We have examined whether the effects of singlet oxygen (1O2) produced by photodynamic action on the mitochondrial permeability transition (PT) can be modulated by the localization of photosensitizers in irradiated mitochondria. We have previously shown that oxidation due to 1O2
Manuel Gutiérrez-Aguilar et al.
Archives of biochemistry and biophysics, 494(2), 184-191 (2009-12-10)
The mitochondrial permeability transition (PT) involves the opening of a mitochondrial unselective channel (MUC) resulting in membrane depolarization and increased permeability to ions. PT has been observed in many, but not all eukaryotic species. In some species, PT has been
Jason M Gooding et al.
European journal of biochemistry, 271(5), 954-961 (2004-03-11)
Recent studies have suggested that parts of the hepatic activities of diacylglycerol acyltransferase and acyl cholesterol acyltransferase are expressed in the lumen of the endoplasmic reticulum (ER). However the ER membrane is impermeable to the long-chain fatty acyl-CoA substrates of
F Agani et al.
Molecular pharmacology, 54(5), 749-754 (1998-11-06)
In response to hypoxia, mammalian cells express multiple gene products [including erythropoietin (EPO) and vascular endothelial growth factor (VEGF)] that serve to increase O2 delivery, as well as glucose transporters and glycolytic enzymes (such as enolase 1) that allow metabolic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service