Skip to Content
Merck
All Photos(1)

Key Documents

M8536

Supelco

2-(Methylamino)propiophenone hydrochloride solution

drug standard, 1.0 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

drug standard

Quality Level

drug control

Home Office Schedule 1; stupéfiant (France); kontrollierte Droge in Deutschland; (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

2-8°C

SMILES string

Cl.CNC(C)C(=O)c1ccccc1

InChI

1S/C10H13NO.ClH/c1-8(11-2)10(12)9-6-4-3-5-7-9;/h3-8,11H,1-2H3;1H

InChI key

LTJXNCHDKYZOSY-UHFFFAOYSA-N

Application

2-(Methylamino)propiophenone hydrochloride may be used as an analytical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Mixture of optical isomers

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic method development and validation for the simultaneous quantitation of naproxen sodium and pseudoephedrine hydrochloride impurities
Ekpe A, et al.
Journal of Chromatographic Science, 39(3), 81-86 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service