Skip to Content
Merck
All Photos(3)

Key Documents

M7304

Sigma-Aldrich

Magnesium chloride hexahydrate

meets USP testing specifications

Synonym(s):

MgCl2 hexahydrate, MgCl2.6H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2 · 6H2O
CAS Number:
Molecular Weight:
203.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

application(s)

pharmaceutical (small molecule)

SMILES string

O.O.O.O.O.O.Cl[Mg]Cl

InChI

1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2

InChI key

DHRRIBDTHFBPNG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Biocompatible topical delivery system of high-molecular-weight hyaluronan into human stratum corneum using magnesium chloride.: This research demonstrates a novel topical delivery system utilizing magnesium chloride hexahydrate to enhance the penetration of high-molecular-weight hyaluronan into the skin, with potential applications in dermatology and wound healing (Fujii et al., 2023).

  • Improving tear ferning patterns collected from goats and camels after adding various electrolyte solutions.: This research focuses on the effects of adding magnesium chloride hexahydrate to tear samples from animals, aiming to enhance the understanding of electrolyte balance and its diagnostic relevance in veterinary medicine (Fagehi et al., 2022).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingchu Gong et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 93, 260-266 (2015-04-25)
Brittleness is one of the important material properties that influences the success or failure of powder compaction. We have discovered that the reciprocal of diametrical elastic strain at fracture is the most suitable tablet brittleness indices (TBIs) for quantifying brittleness
Monica Mazzarino et al.
Analytica chimica acta, 837, 70-82 (2014-07-09)
This article describes a liquid chromatographic/tandem mass spectrometric method, based on the use of precursor ion scan as the acquisition mode, specifically developed to detect indole-derived cannabinoids (phenylacetylindoles, naphthoylindoles and benzoylindoles) in biological fluids (saliva, urine and blood). The method
Liang Yang et al.
International journal of pharmaceutics, 471(1-2), 258-263 (2014-05-20)
Telmisartan (TEL) belongs to BCS class II (low solubility/high permeability) and exhibits the pH-dependent drug release. Since 3-aminopropyl functionalized magnesium phyllosilicate (aminoclay) can intercalate or adsorb the negatively charged molecules via the electrostatic interaction, TEL-aminoclay complex was synthesized to improve
Ikuo Yamamiya et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1485-1492 (2014-07-09)
Tegafur (FT), a prodrug of 5-fluorouracil, is a chiral molecule, a racemate of R- and S-isomers, and CYP2A6 plays an important role in the enantioselective metabolism of FT in human liver microsomes (R-FT > S-FT). This study examined the enantioselective
Heleen Kraan et al.
Vaccine, 33(17), 2030-2037 (2015-03-17)
In the near future oral polio vaccine (OPV) will be replaced by inactivated polio vaccine (IPV) as part of the eradication program of polio. For that reason, there is a need for substantial amount of safe and more affordable IPV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service