Skip to Content
Merck
All Photos(1)

Key Documents

M0950000

DL-Methionine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Beilstein:
636185
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methionine & derivatives

manufacturer/tradename

EDQM

mp

280 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CSCCC(N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ᴅʟ-Methionine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pilar Lloris-Garcerá et al.
Journal of molecular biology, 426(11), 2246-2254 (2014-04-03)
The increasing number of solved membrane protein structures has led to the recognition of a common feature in a large fraction of the small-molecule transporters: inverted repeat structures, formed by two fused homologous membrane domains with opposite orientation in the
Shawn Flanagan et al.
Antimicrobial agents and chemotherapy, 59(1), 178-185 (2014-10-22)
Prolonged treatment with the oxazolidinone linezolid is associated with myelosuppression, lactic acidosis, and neuropathies, toxicities likely caused by impairment of mitochondrial protein synthesis (MPS). To evaluate the potential of the novel oxazolidinone tedizolid to cause similar side effects, nonclinical and
Qian Ren et al.
Nucleic acids research, 42(14), 9366-9382 (2014-07-20)
The dicistrovirus intergenic internal ribosome entry site (IGR IRES) directly recruits the ribosome and initiates translation using a non-AUG codon. A subset of IGR IRESs initiates translation in either of two overlapping open reading frames (ORFs), resulting in expression of
Leen Mathys et al.
PloS one, 9(6), e101181-e101181 (2014-06-27)
N-linked glycans covering the surface of the HIV-1 glycoprotein gp120 are of major importance for the correct folding of this glycoprotein. Of the, on average, 24 N-linked glycans present on gp120, the glycan at Asn260 was reported to be essential
Dylan Thomas et al.
Chembiochem : a European journal of chemical biology, 15(11), 1607-1613 (2014-07-22)
Protein arginine N-methyltransferases (PRMTs) catalyze methyl-group transfer from S-adenosyl-L-methionine onto arginine residues in proteins. In this study, modifications were introduced at the guanidine moiety of a peptidyl arginine residue to investigate how changes to the PRMT substrate can modulate enzyme

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service