Skip to Content
Merck
All Photos(2)

Key Documents

L9774

Sigma-Aldrich

LR white acrylic resins

Medium

Synonym(s):

LR white resin, Acryl resin LR white

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

biological source

synthetic

Quality Level

refractive index

n/D 1.4592-1.4610

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

LR white acrylic resins has been used:
  • to embed microslicer semithin sections of gills
  • to embed left femurs
  • to treat dehydrated samples of buds and flowers for fixation and embedding

Benzoyl peroxide (B5907) is specifically formulated as an initiator for these resins.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative changes in metallothionein expression in target cell-types in the gills of turbot (Scophthalmus maximus) exposed to Cd, Cu, Zn and after a depuration treatment
Alvarado N E, et al.
Aquatic Toxicology (Amsterdam, Netherlands), 77(1), 64-77 (2006)
Ubiquitin E3 ligase Wwp1 negatively regulates osteoblast function by inhibiting osteoblast differentiation and migration
Shu L, et al.
Journal of Bone and Mineral Research, 28(9), 1925-1935 (2013)
LR White Acrylic Resin
Yeung EC and Huang BQ
Plant Microtechniques and Protocols, 103-103 (2015)
Distribution of arabinogalactan proteins during microsporogenesis in the anther of Bellis perennis L.(Asteraceae).
Chudzik B
ACTA BIOLOGICA CRACOVIENSIA Series Botanica, 56, 49-49 (2014)
Mechanisms of desensitization to parathyroid hormone in human osteoblast-like SaOS-2 cells
Fukayama S, et al.
Endocrinology, 131(4), 1757-1769 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service